Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GJD3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2840460.1ML
Description
GJD3 Polyclonal specifically detects GJD3 in Human samples. It is validated for Western Blot.Specifications
GJD3 | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
125111 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Connexin-31.9, Cx30.2, Cx31.9, CX31.9connexin-31.9, Gap junction alpha-11 protein, Gap junction chi-1 protein, gap junction protein, chi 1, 31.9kDa, gap junction protein, delta 3, 31.9kDa, GJA11connexin 31.9, GJC1gap junction delta-3 protein | |
The immunogen is a synthetic peptide directed towards the C-terminal region of GJD3 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction