Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Gje1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582514
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582514 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582514 Supplier Invitrogen™ Supplier No. PA582514
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KSLYLCDARS LGENMIIRCM VPEHFE Highest antigen sequence indentity to the following orthologs: Mouse - 63%, Rat - 63%.

Mediates calcium-independent ATP release, suggesting activity as a hemichannel. Does not form functional gap junctions. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gje1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Gje1
Gene Accession No. A6NN92
Gene Alias AEY12; connexin 23; connexin-23; Cx23; D230044M03Rik; Gap junction epsilon-1 protein; gap junction protein, epsilon 1; Gje1; Gjf1; Gsfaey12; putative gap junction protein connexin Cx43.4
Gene Symbols Gje1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GJE1. Recombinant protein control fragment (Product #RP-91202).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100126572
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.