Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GK2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP156691
Description
GK2 Polyclonal specifically detects GK2 in Human samples. It is validated for Western Blot.Specifications
GK2 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
EC 2.7.1.30, GK 2, GKP2, GKTAATP:glycerol 3-phosphotransferase 2, Glycerokinase 2, glycerol kinase 2, glycerol kinase pseudogene 2, glycerol kinase testis specific 2, Glycerol kinase, testis specific 2 | |
Rabbit | |
Affinity purified | |
RUO | |
2712 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q01415 | |
GK2 | |
Synthetic peptides corresponding to GK2(glycerol kinase 2) The peptide sequence was selected from the C terminal of GK2. Peptide sequence QIQATESEIRYATWKKAVMKSMGWVTSQSPEGGDPSIFSSLPLGFFIVSS. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%; Crab-eating macaque: 92%; Rat: 84%; Mouse: 84%;. | |
Human, Mouse, Equine, Guinea Pig, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction