Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GK5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | GK5 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
GK5 Polyclonal specifically detects GK5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
GK5 | |
Polyclonal | |
Rabbit | |
Protein Kinase | |
ATP:glycerol 3-phosphotransferase 5, EC 2.7.1.30, FLJ33582, FLJ45739, GK 5, Glycerokinase 5, glycerol kinase 5 (putative), MGC40579, putative glycerol kinase 5 | |
GK5 | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
256356 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:QQSAMFGECCFQTGDVKLTMGTGTFLDINTGNSLQQTTGGFYPLIGWKIGQEVVCLAESNAGDTGTAIKWAQQLDLFTDAAET | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title