Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GK5 Monoclonal Antibody (2C11)
GREENER_CHOICE

Catalog No. PIMA523216
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA523216 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA523216 Supplier Invitrogen™ Supplier No. MA523216
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MSGLLTDPEQ RAQEPRYPGF VLGLDVGSSV IRCHVYDRAA RVCGSSVQKV ENLYPQIGWV EIDPDVLWIQ FVAVIKEAVK AAGIQMNQI.

GK5 (Glycerol Kinase 5) is a Protein Coding gene. Diseases associated with GK5 include Type 1 Diabetes Mellitus 7 and Type 1 Diabetes Mellitus 13. Among its related pathways are glycerol degradation and triacylglycerol biosynthesis. Gene Ontology (GO) annotations related to this gene include phosphotransferase activity, alcohol group as acceptor and glycerol kinase activity. An important paralog of this gene is GK. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GK5
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2C11
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene GK5
Gene Accession No. Q6ZS86
Gene Alias ATP:glycerol 3-phosphotransferase 5; AV095337; C330018K18Rik; G630067D24Rik; GK 5; Gk5; glycerokinase 5; glycerol kinase 5 (putative); putative glycerol kinase 5; RGD1311223
Gene Symbols GK5
Host Species Mouse
Immunogen GK5 (NP_689989, 1 a.a. ~ 89 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 256356
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.