Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GK5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560045
Description
Immunogen sequence: NCCFGTIDTW LLYKLTKGSV YATDFSNAST TGLFDPYKMC WSGMITSLIS IPLSLLPPVR DTSHNFGSVD EEIFGVPIPI VAL Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 82%.
GK5 (Glycerol Kinase 5) is a Protein Coding gene. Diseases associated with GK5 include Type 1 Diabetes Mellitus 7 and Type 1 Diabetes Mellitus 13. Among its related pathways are glycerol degradation and triacylglycerol biosynthesis. Gene Ontology (GO) annotations related to this gene include phosphotransferase activity, alcohol group as acceptor and glycerol kinase activity. An important paralog of this gene is GK. [GeneCards].
Specifications
GK5 | |
Polyclonal | |
Unconjugated | |
GK5 | |
ATP:glycerol 3-phosphotransferase 5; AV095337; C330018K18Rik; G630067D24Rik; GK 5; Gk5; glycerokinase 5; glycerol kinase 5 (putative); putative glycerol kinase 5; RGD1311223 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
256356 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZS86 | |
GK5 | |
Recombinant protein corresponding to Human GK5. Recombinant protein control fragment (Product #RP-96444). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction