Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GKAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557159
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557159 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557159 Supplier Invitrogen™ Supplier No. PA557159
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ITQWEAKYKE VKARNAQLLK MLQEGEMKDK AEILLQVDES QSIKNELTIQ VTSLHAALEQ ERSKVKVLQA ELAKYQGGRK GKRNSESDQC R Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

The approximately 142 kDa Tankyrase protein is ubiquitously expressed in mammalian cells and is localized to human telomeres. It has homology to ankyrins and to the catalytic domain of poly (adenosine diphosphate-ribose) polymerase (PARP). Tankyrase binds to TRF1 (telomeric repeat binding factor-1), which negatively regulates the maintenance of telomere length.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GKAP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Gkap1
Gene Accession No. Q5VSY0, Q9JMB0
Gene Alias 42 kD cGMP-dependent protein kinase anchoring protein; 42kDa; 4933400B15Rik; cGMP-dependent protein kinase anchoring protein; cGMP-dependent protein kinase anchoring protein 42kDa; cGMP-dependent protein kinase-anchoring protein of 42 kDa; D13Ertd340e; FKSG21; G kinase anchoring protein 1; G kinase-anchoring protein 1; GKAP1; Gkap42; protein kinase anchoring protein GKAP42
Gene Symbols Gkap1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GKAP1. Recombinant protein control fragment (Product #RP-95675).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56278, 80318
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.