Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GKN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119133
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119133 200 μL
PIPA5121958 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119133 Supplier Invitrogen™ Supplier No. PA5119133
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GKN1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Gastrokine 1, also known as GKN1 and CA11, is a 185 amino acid containing secreted protein belonging to the gastrokine family with a BRICHOS domain and an asn-rich region. GKN1 (also known as an antioncogene) is reported to be down-regulated in human gastric cancer tissue as compared to normal gastric mucosa. Differential display of normal and cancerous gastric tissue suggested that this protein is predominantly expressed in stomach and the expression is down-regulated to considerable frequency in cancer tissue or gastric cancer cell lines examined. This extra cellular protein is co-secreted with mucins and is a signaling growth factor with mitogenic activity and may be involved in maintaining the integrity of the gastric mucosal epithelium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GKN1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene GKN1
Gene Accession No. Q8HYA9, Q9NS71
Gene Alias 18 kDa antrum mucosa protein; 2200002K21Rik; Amp18; AMP-18; Amp18; gastrokine; BRICD1; BRICHOS domain containing 1; Ca11; FOV; foveolin; gastrokine 1; gastrokine-1; Gkn1; Protein CA11; Protein CA11 homolog; UNQ489/PRO1005
Gene Symbols GKN1
Host Species Rabbit
Immunogen Recombinant protein Gastrokine 1. The antigen corresponds to amino acid range Ala22-Asn199 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100502561, 297418, 56287
Target Species Human, Rat, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.