Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GKN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588988
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588988 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588988 Supplier Invitrogen™ Supplier No. PA588988
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LGVFLAPALA NYNINVNDDN NNAGSGQQSV SVNNEHNVAN VDNNNGWDSW NSIWDYGNGF AATRLFQKKT CIVHKMNKEV MPSIQSLDAL VKEKKLQGKG PGGPPPKGLM YSVNPNKVDD LSKFGKNIAN MCRGIPTYMA EEMQEASLFF YSGTCYTTSV L; Positive Samples: Mouse stomach.

Gastrokine 1, also known as GKN1 and CA11, is a 185 amino acid containing secreted protein belonging to the gastrokine family with a BRICHOS domain and an asn-rich region. GKN1 (also known as an antioncogene) is reported to be down-regulated in human gastric cancer tissue as compared to normal gastric mucosa. Differential display of normal and cancerous gastric tissue suggested that this protein is predominantly expressed in stomach and the expression is down-regulated to considerable frequency in cancer tissue or gastric cancer cell lines examined. This extra cellular protein is co-secreted with mucins and is a signaling growth factor with mitogenic activity and may be involved in maintaining the integrity of the gastric mucosal epithelium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GKN1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GKN1
Gene Accession No. Q9CR36
Gene Alias 18 kDa antrum mucosa protein; 2200002K21Rik; Amp18; AMP-18; Amp18; gastrokine; BRICD1; BRICHOS domain containing 1; Ca11; FOV; foveolin; gastrokine 1; gastrokine-1; Gkn1; Protein CA11; Protein CA11 homolog; UNQ489/PRO1005
Gene Symbols GKN1
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66283
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.