Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GKN2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA545701
Description
Peptide sequence: NIISPSNNGG NVQETVTIDN EKNTAIINIH AGSCSSTTIF DYKHGYIASR Sequence homology: Dog: 85%; Horse: 91%; Human: 100%; Mouse: 85%; Rabbit: 77%; Rat: 92%.
The function of this protein remains unknown.
Specifications
GKN2 | |
Polyclonal | |
Unconjugated | |
GKN2 | |
1810036H07Rik; Blot; Blottin; BRICD1B; BRICHOS domain containing 1B; Down-regulated in gastric cancer; down-regulated in gastric cancer GDDR; gastrokine 2; gastrokine-2; GDDR; Gkn2; PRO813; RGD1311934; TFIZ1; trefoil factor interactions(z) 1; UNQ465/PRO813; VLTI465 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
200504 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86XP6 | |
GKN2 | |
Synthetic peptide directed towards the N-terminal of human GKN2 (aa 25-74). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction