Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GKN3P Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549018
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549018 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549018 Supplier Invitrogen™ Supplier No. PA549018
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GKN3P Polyclonal Antibody for Western Blot

May inhibit gastric epithelial cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GKN3P
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GKN3P
Gene Accession No. P0CG01
Gene Alias Gastrokine-3; GKN3P
Gene Symbols GKN3P
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 108-134 amino acids from the Central region of human GKN3P.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100132413
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.