Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GL54D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557701
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557701 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557701 Supplier Invitrogen™ Supplier No. PA557701
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FQVKVCDAGE DLRNCMKRKK QIRIQYKPSL Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 43%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen GL54D
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MGAT4D
Gene Accession No. A6NG13
Gene Alias alpha-1,3-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase-like protein MGAT4D; GlcNAcT-I inhibitory protein; glycosyltransferase 54 domain containing 1; glycosyltransferase 54 domain-containing protein; GnT1IP; mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase family, member D; MGAT4 family member D; MGAT4 family, member D; MGAT4D; MGAT4D (HGNC:43619); N-acetylglucosaminyltransferase MGAT1 inhibitory protein
Gene Symbols MGAT4D
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GL54D. Recombinant protein control fragment (Product #RP-96758).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152586
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.