Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GL7 (T and B Cell Activation Marker) Antibody (GL-7), Novus Biologicals™
SDP

Catalog No. p-5324096 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP20036210 100 μg
NBP20036255 500 μg
NB018829 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP20036210 Supplier Novus Biologicals Supplier No. NBP200362100UG
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

GL7 (T and B Cell Activation Marker) Monoclonal specifically detects GL7 (T and B Cell Activation Marker) in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.

Specifications

Antigen GL7 (T and B Cell Activation Marker)
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone GL-7
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry < = 0.5 ug/10^6 cells in 100 ul, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation PBS, pH 7.2 with 0.09% Sodium Azide
Host Species Rat
Immunogen The immunogen for this antibody is T- and B-cell activation antigen (GL7, Ly-77).
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.