Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scientific Cell Company Glass Filter for Testing Wavelength Accuracy

Catalog No. 14397501
Click to view available options
Type:
Glass filter

Includes

DAkkS calibration certificate

For testing the wavelength accuracy.

  • Certified at wavelengths: 279, 361, 453, 536, 638nm slit width
  • 1nm serial no.
  • Type: Solid
  • Individual filter
  • Light Path: Holmium Oxide
  • Packed in high-quality storage case

Specifications

Type Glass filter
For Use With (Application) Testing the wavelength accuracy
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.