Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Glass Jointed Powder Funnels
SCI_ED

Catalog No. p-7147170
Click to view available options
Diameter (Metric)Top:
45 mm
55 mm
70 mm
100 mm
Joint Number:
14/23
29/32

Ideal for transferring powder reagents between pieces of labware. Eisco™ Glass Jointed Powder Funnels feature ground joints to ensure tight connections with other pieces of glassware.

  • Made from high quality borosilicate 3.3 glass
  • Designed with ground joints to ensure tight connections with other pieces of glassware
  • Ideal for transferring powder reagents
  • 60° angle stem
  • Autoclavable

Warranty and Services

1 year limited manufacturer warranty

Specifications

Diameter (Metric)Top 45 mm
Material Borosilicate Glass
Height (Metric) 12.7 cm
Angle 60°
Autoclavable Autoclavable
Color Clear
Description Plain stem
Graduated No
For Use With (Application) Transferring liquids
Joint Number 14/23
Length (Metric) Stem 40.64 mm
Quantity 1
Length (English) Overall 3.25 in.
Height (English) 5 in.
No. of Pores 1
Product Type Filling Funnel
Material (Membrane) Glass
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.