Learn More
Eisco™ Glass Rectangular Block
SCI_ED
Catalog No. S24378 Shop All Eisco Products
Change view
Click to view available options
Dimensions (L x W x H):
100 x 60 x 18 mm
115 x 65 x 18 mm
125 x 65 x 18 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
S24378 100 x 60 x 18 mm
S24379 115 x 65 x 18 mm
S24380 125 x 65 x 18 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S24378 Supplier Eisco™ Supplier No. PH0566C
Add to Cart
Add to Cart
With all angles accurately worked and optically worked surfaces.
  • Made from imported optical glass
  • Glass free from bubbles
Specifications
Product Type Index of Refraction Plate
Lens Material Glass
For Use With (Application) Studying refraction and geometrical optics in physics and physical science classes
Dimensions (L x W x H) 100 x 60 x 18 mm