Learn More
United Scientific Supplies Glass Separatory Funnel
SDPSCI_ED
Catalog No. S82837
Click to view available options
Capacity (Metric):
50 mL
100 mL
125 mL
250 mL
500 mL
1000 mL
2000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy
Glass Separatory Funnels are pear shaped for easy handling and use.
  • These pear-shaped borosilicate glass funnels include a PTFE stopcock and a plastic stopper.
  • Feature a marking spot and large white block letters for easy identification of capacity, as per ASTM E1096.
  • PTFE stopcocks are compliant with ASTM standard E675.
Specifications
Capacity (Metric) 1000 mL
Material Borosilicate Glass 3.3
Color Clear
Quantity 1 Ea.
Product Type Separatory Funnel
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.