Learn More
Labconco™ Glassware Washer Dispenser
GSA_VASDP
Accessories for use with Labconco Glassware Washers with CleanWorks OS.
$1440.00 - $2950.00
Specifications
Material Plastic
Product Type Washer Dispenser
For Use With (Equipment) Labconco FlaskScrubber or SteamScrubber
Quantity 1/Ea.
View More Specs
Products 3
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
15-352-020
GSA_VASDP
View Documents
Labconco™
4587520
Liquid Detergent Dispenser
Each for $1,451.00
Only null left
Add to Cart
15-352-021
GSA_VASDP
View Documents
Labconco™
4595120
Acid Rinse Dispenser
Each for $1,440.00
Only null left
Add to Cart
15-352-022
GSA_VASDP
View Documents
Labconco™
4679700
Detergent & Acid Rinse Dispenser
Each for $2,950.00
Only null left
Add to Cart
Description
Description
  • Racks and accessories for use with Labconco FlaskScrubber, SteamScrubber, FlaskScrubber Vantage Glassware Washers.
  • Liquid Detergent Dispenser connects to any SteamScrubber or FlaskScrubber Washer to automate liquid detergent dispensing. Includes peristaltic pump, float switch, tubing, and one-gallon plastic container. Detergent not included.
  • Acid Rinse Dispenser connects to any SteamScrubber or FlaskScrubber Washer to automate acid neutralizing rinse aid dispensing. Includes peristaltic pump, float switch, tubing, and a one-liter plastic container. Rinse aid not included.
  • Detergent & Acid Rinse Dispenser connects to any SteamScrubber or FlaskScrubber Washer to automate liquid detergent & acid rinse aid dispensing on washers with 9-digit catalog number. Includes 1 gal detergent & 1 L rinse aid containers. Detergent & rinse aid not included.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Plastic
Washer Dispenser
Labconco FlaskScrubber or SteamScrubber
1/Ea.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.