Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glassy carbon spherical powder, 0.4-12 micron, type 2

Catalog No. AA3800809
Click to view available options
Quantity:
10 g
50 g
250 g

Catalog No. AA3800809

Supplier: Thermo Scientific Chemicals 03800809

Only null left
Add to Cart

In typical measurements, 8 cm downstream of the burner, of species profiles versus λ for a flame doped with type 1 glassy carbon spherical powder, as parts per million, normalized to the Ar36 peak area and the measured particle cross sectional area in Computer Model Study of Carbon Combustion in Low Pressure Laminar H 2+ O 2 Flames. Glassy carbon spherical powder (10-20 μm diameter) modified with cysteine methyl ester is found to be an inexpensive, novel material for the rapid removal of large quantities of toxic heavy metal ions such as Cd(II), Cu(II) and As(III) from aqueous media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00133992

Specifications

Packaging Glass bottle
Quantity 10 g
UN Number UN1325
Solubility Information Insoluble in water.
Chemical Name or Material Glassy carbon spherical powder
EINECSNumber 231-153-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.