Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLAST Recombinant Rabbit Monoclonal Antibody (JA30-35)
GREENER_CHOICE

Catalog No. PIMA541231
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541231 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA541231 Supplier Invitrogen™ Supplier No. MA541231
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

GLAST Recombinant Monoclonal Antibody for IHC (F), IHC (P), Western Blot

This gene encodes a member of a member of a high affinity glutamate transporter family. Mutations in this gene are associated with episodic ataxia, Type 6. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLAST
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JA30-35
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene SLC1A3
Gene Accession No. P24942, P43003, P56564
Gene Alias AI504299; B430115D02Rik; EA6; Eaat1; EAAT-1; Excitatory amino acid transporter 1; FLJ25094; GLAST; GLAST1; GLAST-1; Glial glutamate transporter; glial high affinity glutamate transporter; GLU-T; GluT-1; glutamate transporter; glutamate/aspartate transporter; Gmt1; High-affinity neuronal glutamate transporter; MGluT1; Slc1a3; Sodium-dependent glutamate/aspartate transporter 1; solute carrier family 1 (glial high affinity glutamate transporter), member 3; solute carrier family 1 member 3; solute carrier family 1, member 3
Gene Symbols SLC1A3
Host Species Rabbit
Immunogen Synthetic peptide within Human GLAST aa 171-220.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20512, 29483, 6507
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.