Learn More
GLB1 Mouse anti-Canine, Human, Clone: OTI1C9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505544

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
GLB1 cleaves beta-linked terminal galactosyl residues from gangliosides, glycoproteins, and glycosaminoglycans. Isoform 2 has no beta-galactosidase catalytic activity, but plays functional roles in the formation of extracellular elastic fibers (elastogenesis) and in the development of connective tissue. This protein seems to be identical to the elastin-binding protein (EBP), a major component of the non-integrin cell surface receptor expressed on fibroblasts, smooth muscle cells, chondroblasts, leukocytes, and certain cancer cell types. In elastin producing cells, associates with tropoelastin intracellularly and functions as a recycling molecular chaperone which facilitates the secretions of tropoelastin and its assembly into elastic fibers.Specifications
GLB1 | |
Monoclonal | |
Unconjugated | |
GLB1 | |
EBP, ELNR1, MPS4B | |
Mouse | |
Affinity Chromatography | |
RUO | |
2720, 403873 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1C9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P16278, Q9TRY9 | |
GLB1 | |
Full length human recombit protein of human GLB1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Canine, Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.