Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLB1 Mouse anti-Dog, Human, Monkey, Mouse, Rat, Clone: 10B2, Abnova™

Catalog No. 89934200 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-200 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-200 Supplier Abnova Corporation Supplier No. MAB12856
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full length recombinant human GLB1.

This gene encodes beta-galactosidase-1, a lysosomal enzyme that hydrolyzes the terminal beta-galactose from ganglioside substrates and other glycoconjugates. Defects in this gene are the cause of GM1-gangliosidosis and Morquio B syndrome. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Antigen GLB1
Applications Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 10B2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant human GLB1.
Dilution Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:200-1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.3 (50% glycerol, 1% BSA, 0.02% sodium azide).
Gene Alias EBP/ELNR1
Gene Symbols GLB1
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human GLB1.
Purification Method Protein A/G purification
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2720
Target Species Canine, Human, Monkey, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.