Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GLB1L2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563023
Description
Immunogen sequence: RGRVNYGENI DDQRKGLIGN LYLNDSPLKN FRIYSLDMKK SFFQRFGLDK WSSL Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 80%.

Specifications
GLB1L2 | |
Polyclonal | |
Unconjugated | |
GLB1L2 | |
Beta-galactosidase-1-like protein 2; galactosidase beta 1 like 2; galactosidase beta 1-like 2; galactosidase, beta 1-like 2; GLB1L2; MST114; MSTP014; MSTP114; UNQ210/PRO236 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
89944 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IW92 | |
GLB1L2 | |
Recombinant protein corresponding to Human GLB1L2. Recombinant protein control fragment (Product #RP-101367). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction