Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLB1L3 Polyclonal Antibody
GLB1L3 Polyclonal Antibody
Supplier: Thermo Scientific PA558983
Description
GLB1L3 Polyclonal Antibody for IHC

Specifications
GLB1L3 | |
Polyclonal | |
Unconjugated | |
GLB1L3 | |
4921509F24Rik; beta-galactosidase-1-like protein 3; galactosidase beta 1 like 3; galactosidase beta 1-like 3; galactosidase, beta 1 like 3; galactosidase, beta 1-like 3; GLB1L3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
112937 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NCI6 | |
GLB1L3 | |
Recombinant protein corresponding to Human GLB1L3. Recombinant protein control fragment (Product #RP-97573). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction