Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLCCI1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988540
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8540 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8540 Supplier Bioss Supplier No. BS8410R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GLCCI1 is a protein coding gene. This gene encodes a protein of unknown function. Expression of this gene is induced by glucocorticoids and may be an early marker for glucocorticoid-induced apoptosis. Single nucleotide polymorphisms in this gene are associated with a decreased response to inhaled glucocorticoids in asthmatic patients.

Specifications

Antigen GLCCI1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GLCCI1
Gene Accession No. Q86VQ1
Gene Alias 2310047L21Rik; A130036A18Rik; Fam117c; GCTR; Gig18; Glcci1; glucocorticoid induced 1; glucocorticoid induced transcript 1; Glucocorticoid-induced gene 18 protein; Glucocorticoid-induced transcript 1 protein; RGD1563612; Testhymin; Thymocyte/spermatocyte selection protein 1; Tssn1
Gene Symbols GLCCI1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GLCCI1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113263
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.