Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLCE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559194
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559194 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559194 Supplier Invitrogen™ Supplier No. PA559194
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SDKAIQFPRR SSSGFRVDGF EKRAAASESN NYMNHVAKQQ SEEAFPQEQQ KAPPVVGGFN SNVGSKVLGL KYEEIDCLIN DEHTIKGRRE GNEVFLPFTW VEKYFDVYGK VVQYDGYDR Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.

Converts D-glucuronic acid residues adjacent to N-sulfate sugar residues to L-iduronic acid residues, both in maturing heparan sulfate (HS) and heparin chains. This is important for further modifications that determine the specificity of interactions between these glycosaminoglycans and proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLCE
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GLCE
Gene Accession No. O94923
Gene Alias 1110017N23Rik; AI747411; C130034A12Rik; D-glucuronyl C5-epimerase; GLCE; glucuronic acid epimerase; glucuronyl C5-epimerase; heparan sulfate C5-epimerase; heparan sulfate epimerase; heparan sulfate-glucuronic acid C5-epimerase; heparin sulfate C5-epimerase; heparin/heparan sulfate:glucuronic acid C5-epimerase; heparin/heparan sulfate-glucuronic acid C5-epimerase; Heparosan-N-sulfate-glucuronate 5-epimerase; HSEPI; KIAA0836; RGD1565253; UDP-glucuronic acid epimerase
Gene Symbols GLCE
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GLCE. Recombinant protein control fragment (Product #RP-98073).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26035
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.