Learn More
Invitrogen™ GLCE Polyclonal Antibody

Description
Immunogen sequence: ETSEGVSLQL GNTKDFIISF DLKFLTNGSV SVVLETTEKN QLFTIHYVSN AQLIAFKERD IYYGIGPRTS WSTVTRDLVT DLRKGVGLSN TKAVKPTKIM PKKVVRLI Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.
Specifications
Specifications
| Antigen | GLCE |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | GLCE |
| Gene Accession No. | O94923 |
| Gene Alias | 1110017N23Rik; AI747411; C130034A12Rik; D-glucuronyl C5-epimerase; GLCE; glucuronic acid epimerase; glucuronyl C5-epimerase; heparan sulfate C5-epimerase; heparan sulfate epimerase; heparan sulfate-glucuronic acid C5-epimerase; heparin sulfate C5-epimerase; heparin/heparan sulfate:glucuronic acid C5-epimerase; heparin/heparan sulfate-glucuronic acid C5-epimerase; Heparosan-N-sulfate-glucuronate 5-epimerase; HSEPI; KIAA0836; RGD1565253; UDP-glucuronic acid epimerase |
| Gene Symbols | GLCE |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.