Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565876
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565876 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565876 Supplier Invitrogen™ Supplier No. PA565876
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HSPHQEPTVVN QIVPLSGERR YSMPPLFHTH YVPDIVRCVP PFREIAFLEP REITLPEAKD KLSQQILELF ETCQQQISDL KKKELCRT Highest antigen sequence identity to the following orthologs - mouse 89%, rat 89%.

GLD2 is cytoplasmic poly(A) RNA polymerase that adds successive AMP monomers to the 3'-end of specific RNAs, forming a poly(A) tail. In contrast to the canonical nuclear poly(A) RNA polymerase, it only adds poly(A) to selected cytoplasmic mRNAs. GLD2 does not play a role in replication-dependent histone mRNA degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLD2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Papd4
Gene Accession No. Q6PIY7
Gene Alias 8030446C20Rik; AI649009; cytoplasmic poly(A) polymerase; Gld2; GLD-2; hGLD-2; hypothetical protein LOC553626; MGC138008 protein; mGLD-2; Palp4; PAP associated domain containing 4; PAP-associated domain-containing protein 4; PAPD4; poly(A) RNA polymerase D4, non-canonical; Poly(A) RNA polymerase GLD2; Tent2; Terminal nucleotidyltransferase 2; terminal uridylyltransferase 2; TUT2; TUTase 2; TUTase2; zgc:110560
Gene Symbols Papd4
Host Species Rabbit
Immunogen Recombinant Human GLD2. Recombinant protein control fragment (Product #RP-104999).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 167153
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.