Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569570
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569570 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569570 Supplier Invitrogen™ Supplier No. PA569570
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 83%.

GLD2 is cytoplasmic poly(A) RNA polymerase that adds successive AMP monomers to the 3'-end of specific RNAs, forming a poly(A) tail. In contrast to the canonical nuclear poly(A) RNA polymerase, it only adds poly(A) to selected cytoplasmic mRNAs. GLD2 does not play a role in replication-dependent histone mRNA degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLD2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Papd4
Gene Accession No. Q5U315
Gene Alias 8030446C20Rik; AI649009; cytoplasmic poly(A) polymerase; Gld2; GLD-2; hGLD-2; hypothetical protein LOC553626; MGC138008 protein; mGLD-2; Palp4; PAP associated domain containing 4; PAP-associated domain-containing protein 4; PAPD4; poly(A) RNA polymerase D4, non-canonical; Poly(A) RNA polymerase GLD2; Tent2; Terminal nucleotidyltransferase 2; terminal uridylyltransferase 2; TUT2; TUTase 2; TUTase2; zgc:110560
Gene Symbols Papd4
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of rat GLD2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361878
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.