Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589106
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589106 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589106 Supplier Invitrogen™ Supplier No. PA589106
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MFPNSILGRP PFTPNHQQHN NFFTLSPTVY SHQQLIDAQF NFQNADLSRA VSLQQLTYGN VSPIQTSASP LFRGRKRLSD EKNLPLDGKR QRFHSPHQEP TVVNQIVPLS GERRYSMPPL FHTHYVPDIV RCVPPFREIA FLEPREITLP EAKDKLSQQI LELFETCQQQ ISDLKKKELC RTQLQREIQL LFPQSRLFLV GSSLNGFGTR SSDGDLCLVV KEEPCFFQVN QKTEARHILT LVHKHFCTRL SGYIERPQLI RAKVPIVKFR DKVSCVEFDL NVNNIVGIRN TFLLRTYAYL; Positive Samples: U-87MG, BxPC-3, Mouse heart, Rat heart; Cellular Location: Cytoplasm, Nucleus.

GLD2 is cytoplasmic poly(A) RNA polymerase that adds successive AMP monomers to the 3'-end of specific RNAs, forming a poly(A) tail. In contrast to the canonical nuclear poly(A) RNA polymerase, it only adds poly(A) to selected cytoplasmic mRNAs. GLD2 does not play a role in replication-dependent histone mRNA degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLD2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.88 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Papd4
Gene Accession No. Q5U315, Q6PIY7, Q91YI6
Gene Alias 8030446C20Rik; AI649009; cytoplasmic poly(A) polymerase; Gld2; GLD-2; hGLD-2; hypothetical protein LOC553626; MGC138008 protein; mGLD-2; Palp4; PAP associated domain containing 4; PAP-associated domain-containing protein 4; PAPD4; poly(A) RNA polymerase D4, non-canonical; Poly(A) RNA polymerase GLD2; Tent2; Terminal nucleotidyltransferase 2; terminal uridylyltransferase 2; TUT2; TUTase 2; TUTase2; zgc:110560
Gene Symbols Papd4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human PAPD4 (NP_7761582).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100715, 167153, 361878
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.