Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLD2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA525015

Catalog No. PIPA525015


Only null left
Add to Cart

Description

Description

This antibody is predicted to react with bovine based on sequence homology.

GLD2 is cytoplasmic poly(A) RNA polymerase that adds successive AMP monomers to the 3'-end of specific RNAs, forming a poly(A) tail. In contrast to the canonical nuclear poly(A) RNA polymerase, it only adds poly(A) to selected cytoplasmic mRNAs. GLD2 does not play a role in replication-dependent histone mRNA degradation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GLD2
Polyclonal
Unconjugated
Papd4
8030446C20Rik; AI649009; cytoplasmic poly(A) polymerase; Gld2; GLD-2; hGLD-2; hypothetical protein LOC553626; MGC138008 protein; mGLD-2; Palp4; PAP associated domain containing 4; PAP-associated domain-containing protein 4; PAPD4; poly(A) RNA polymerase D4, non-canonical; Poly(A) RNA polymerase GLD2; Tent2; Terminal nucleotidyltransferase 2; terminal uridylyltransferase 2; TUT2; TUTase 2; TUTase2; zgc:110560
Rabbit
Antigen affinity chromatography, Protein A
RUO
100715, 167153
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.45 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q6PIY7, Q91YI6
Papd4
KLH conjugated synthetic peptide between 58-87 amino acids from the N-terminal region of human GLD2.
400 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.