Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLDC Monoclonal Antibody (3D3D3)
GREENER_CHOICE

Catalog No. PIMA549279
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549279 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549279 Supplier Invitrogen™ Supplier No. MA549279
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human HCCP tissue, human 293T whole cell, human A549 whole cell, rat liver tissue, rat kidney tissue, mouse liver tissue, mouse kidney tissue. IHC: human placenta tissue, human thyroiditis tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Degradation of glycine is brought about by the glycine cleavage system, which is composed of four mitochondrial protein components: P protein (a pyridoxal phosphate-dependent glycine decarboxylase), H protein (a lipoic acid-containing protein), T protein (a tetrahydrofolate-requiring enzyme), and L protein (a lipoamide dehydrogenase). The protein is the P protein, which binds to glycine and enables the methylamine group from glycine to be transferred to the T protein. Defects in this gene are a cause of nonketotic hyperglycinemia (NKH).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLDC
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3D3D3
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene GLDC
Gene Accession No. P23378, Q91W43
Gene Alias D030049L12Rik; D19Wsu57e; GCE; GCSP; GLDC; Glycine cleavage system P protein; glycine cleavage system protein P; glycine decarboxylase; glycine decarboxylase P-protein; glycine decarboxylase, glycine cleavage system protein P); glycine dehydrogenase (aminomethyl-transferring); glycine dehydrogenase (decarboxylating); glycine dehydrogenase (decarboxylating), mitochondrial; glycine dehydrogenase (decarboxylating; glycine decarboxylase, glycine cleavage system protein P); glycine dehydrogenase [decarboxylating], mitochondrial; HYGN1; RP11-472F14.1
Gene Symbols GLDC
Host Species Mouse
Immunogen E.coli-derived human Glycine decarboxylase/GLDC recombinant protein (Position: K574-S1020).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104174, 2731, 309312
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.