Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GLDC Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA526504
Description
GLDC Polyclonal Antibody for Western Blot
Degradation of glycine is brought about by the glycine cleavage system, which is composed of four mitochondrial protein components: P protein (a pyridoxal phosphate-dependent glycine decarboxylase), H protein (a lipoic acid-containing protein), T protein (a tetrahydrofolate-requiring enzyme), and L protein (a lipoamide dehydrogenase). The protein is the P protein, which binds to glycine and enables the methylamine group from glycine to be transferred to the T protein. Defects in this gene are a cause of nonketotic hyperglycinemia (NKH).
Specifications
GLDC | |
Polyclonal | |
Unconjugated | |
GLDC | |
D030049L12Rik; D19Wsu57e; GCE; GCSP; GLDC; Glycine cleavage system P protein; glycine cleavage system protein P; glycine decarboxylase; glycine decarboxylase P-protein; glycine decarboxylase, glycine cleavage system protein P); glycine dehydrogenase (aminomethyl-transferring); glycine dehydrogenase (decarboxylating); glycine dehydrogenase (decarboxylating), mitochondrial; glycine dehydrogenase (decarboxylating; glycine decarboxylase, glycine cleavage system protein P); glycine dehydrogenase [decarboxylating], mitochondrial; HYGN1; RP11-472F14.1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
104174, 2731 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P23378, Q91W43 | |
GLDC | |
KLH conjugated synthetic peptide between 49-77 amino acids from the N-terminal region of human GLDC. | |
400 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction