Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLDN Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501735246 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5246 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5246 Supplier Proteintech Group Inc Supplier No. 261851AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CRG-L2 is a protein that contains olfactomedin-like and collagen-like domains. The encoded protein, which exists in both transmembrane and secreted forms, promotes formation of the nodes of Ranvier in the peripheral nervous system. Mutations in this gene cause a form of lethal congenital contracture syndrome in human patients. Autoantibodies to the encoded protein have been identified in sera form patients with multifocal motor neuropathy.

Specifications

Antigen GLDN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene GLDN
Gene Accession No. Q6ZMI3, Q8BMF8
Gene Alias CLOM, COLM, CRG L2, CRGL2, GLDN, gliomedin, UNC 112
Gene Symbols GLDN
Host Species Rabbit
Immunogen GLDN Fusion Protein Ag24247
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 235379, 342035
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.