Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GLE1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583571
Description
Immunogen sequence: DESQHTESMV LQSSRGIKVE GCVRMYELVH RMKGTEGLRL WQEEQERKVQ ALSEMASEQL KRFDEWKELK QHKEFQDLRE VMEKSSREAL GHQEKLKAEH R.
The proper expression of gene products in eukaryotic cells relies on efficient transport of mRNA molecules out of the nucleus. Gle1 is an essential mRNA export factor in both human and yeast cells. It associates with the nuclear pore complex (NPC) through hCG1 and NUP155 in mammalian cells and in conjunction with inositol hexakisphosphate (IP6), stimulates Dbp5, a member of the DEAD-box helicase family, triggering mRNP remodeling and facilitating RNA export from the nucleus. Recent evidence suggests that mutations in Gle1 causing defects in mRNA export can result in human disease. At least three isoforms of Gle1 are known to exist.
Specifications
GLE1 | |
Polyclonal | |
Unconjugated | |
GLE1 | |
4933405K21Rik; AA553313; GLE1; GLE1 RNA export mediator; GLE1 RNA export mediator (yeast); GLE1 RNA export mediator homolog; GLE1 RNA export mediator-like (yeast; GLE1, RNA export mediator; GLE1L; GLE1-like protein; GLE1-like, RNA export mediator; hGLE1; LCCS; LCCS1; Nucleoporin GLE1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2733 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q53GS7 | |
GLE1 | |
Recombinant protein corresponding to Human GLE1. Recombinant protein control fragment (Product #RP-98406). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction