Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589077
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589077 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589077 Supplier Invitrogen™ Supplier No. PA589077
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPSEGRCWET LKALRSSDKG RLCYYRDWLL RREDVLEECM SLPKLSSYSG WVVEHVLPHM QENQPLSETS PSSTSASALD QPSFVPKSPD ASSAFSPASP ATPNGTKGKD ESQHTESMVL QSSRGIKVEG CVRMYELVHR MKGTEGLRLW QEEQERKVQA LSEMASEQLK RFDEWKELKQ HKEFQDLREV MEKSSREALG HQEKLKAEHR HRAKILNLKL REAEQQRVKQ AEQERLRKEE GQIRLRALYA LQEEMLQLSQ QLDASEQHKA LLKVDLAAFQ TRGNQLCSLI SGIIRASSES; Positive Samples: Jurkat; Cellular Location: Cytoplasm, Cytoplasm, Nucleus, nuclear pore complex.

The proper expression of gene products in eukaryotic cells relies on efficient transport of mRNA molecules out of the nucleus. Gle1 is an essential mRNA export factor in both human and yeast cells. It associates with the nuclear pore complex (NPC) through hCG1 and NUP155 in mammalian cells and in conjunction with inositol hexakisphosphate (IP6), stimulates Dbp5, a member of the DEAD-box helicase family, triggering mRNP remodeling and facilitating RNA export from the nucleus. Recent evidence suggests that mutations in Gle1 causing defects in mRNA export can result in human disease. At least three isoforms of Gle1 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLE1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GLE1
Gene Accession No. Q53GS7
Gene Alias 4933405K21Rik; AA553313; GLE1; GLE1 RNA export mediator; GLE1 RNA export mediator (yeast); GLE1 RNA export mediator homolog; GLE1 RNA export mediator-like (yeast; GLE1, RNA export mediator; GLE1L; GLE1-like protein; GLE1-like, RNA export mediator; hGLE1; LCCS; LCCS1; Nucleoporin GLE1
Gene Symbols GLE1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human GLE1 (NP_0010037221).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2733
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.