Learn More
GLE1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA521816
Description
PA5-21816 targets GLE1 in ICC, IF, IHC (P), and WB applications and shows reactivity with Human samples. The PA5-21816 immunogen is recombit protein fragment contain a sequence corresponding to a region within amino acids 414 and 646 (Q53GS7) of GLE1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications
GLE1 | |
Polyclonal | |
Unconjugated | |
Q53GS7 | |
GLE1 | |
Recombit protein fragment contain a sequence corresponding to a region within amino acids 414 and 646 (Q53GS7) of GLE1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
GLE1 | |
GLE1L, LCCS, LCCS1, hGLE1, GLE1-like protein, GLE1-like, RNA export mediator, nucleoporin GLE1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2733 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.