Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582407
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582407 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582407 Supplier Invitrogen™ Supplier No. PA582407
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LDYRLDPQLQ LHCSDEISSL CAEEAAAQEQ TGQVEECLKV NLLKIKTELC KKEVLNMLKE SKADIFVDPV LHTACALDIK HHCAAITPGR GRQMSCLMEA LEDKRVRLQP ECKKRLNDRI EMWSYAAKVA PADGFSDLAM QVMTSPSKNY Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 98%.

GLG1 is a protein coding gene. Binds fibroblast growth factor and E-selectin (cell-adhesion lectin on endothelial cells mediating the binding of neutrophils).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLG1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GLG1
Gene Accession No. Q92896
Gene Alias AI593353; AW537898; CFR; CFR1; CFR-1; Cysteine-rich fibroblast growth factor receptor; E-selectin ligand 1; ESL1; ESL-1; Glg1; Golgi apparatus protein 1; golgi glycoprotein 1; golgi sialoglycoprotein MG-160; MG160; MG-160; selectin, endothelial cell, ligand; Selel
Gene Symbols GLG1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GLG1. Recombinant protein control fragment (Product #RP-90019).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2734
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.