Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588300
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588300 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588300 Supplier Invitrogen™ Supplier No. PA588300
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RYADQRLSSD CEDQIRIIIQ ESALDYRLDP QLQLHCSDEI SSLCAEEAAA QEQTGQVEEC LKVNLLKIKT ELCKKEVLNM LKESKADIFV DPVLHTACAL DIKHHCAAIT PGRGRQMSCL MEALEDKRVR LQPECKKRLN DRIEMWSYAA KVAPADGFSD LAMQVMTSPS KNYILSVISG SICILFLIGL MCGRITKRVT RELKDRLQYR SETMAYKGLV WSQDVTGSPA; Positive Samples: A-549, LO2, Mouse brain, Rat brain; Cellular Location: Golgi apparatus membrane, Single-pass type I membrane protein.

GLG1 is a protein coding gene. Binds fibroblast growth factor and E-selectin (cell-adhesion lectin on endothelial cells mediating the binding of neutrophils).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLG1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GLG1
Gene Accession No. Q61543, Q92896
Gene Alias AI593353; AW537898; CFR; CFR1; CFR-1; Cysteine-rich fibroblast growth factor receptor; E-selectin ligand 1; ESL1; ESL-1; Glg1; Golgi apparatus protein 1; golgi glycoprotein 1; golgi sialoglycoprotein MG-160; MG160; MG-160; selectin, endothelial cell, ligand; Selel
Gene Symbols GLG1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 974-1203 of human GLG1 (NP_0363332).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20340, 2734
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.