Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLI-1 Antibody (1B9F8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP252475
Description
GLI-1 Monoclonal specifically detects GLI-1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
GLI-1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
GLI family zinc finger 1, Glioma-associated oncogene, glioma-associated oncogene family zinc finger 1, glioma-associated oncogene homolog 1 (zinc finger protein), GLIzinc finger protein GLI1, Oncogene GLI | |
Mouse | |
118 kDa | |
0.1 mg | |
Cancer, Signal Transduction, Stem Cell Signaling Pathway, Transcription Factors and Regulators | |
2735 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1B9F8 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
GLI1 | |
Purified recombinant fragment of human GLI1 (AA: 284-449) expressed in E. Coli. | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction