Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLI-1 Antibody (OTI4E2), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270831H
Description
GLI-1 Monoclonal specifically detects GLI-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Knockdown Validated.Specifications
GLI-1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
GLI1 | |
Recombinant protein fragment corresponding to amino acids 755-1106 of human GLI-1 (NP_005260) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry, KnockDown | |
OTI4E2 | |
Western Blot, Immunohistochemistry, KnockDown Validated | |
GLI family zinc finger 1, Glioma-associated oncogene, glioma-associated oncogene family zinc finger 1, glioma-associated oncogene homolog 1 (zinc finger protein), GLIzinc finger protein GLI1, Oncogene GLI | |
Mouse | |
Affinity Purified | |
Cancer, Signal Transduction, Stem Cell Signaling Pathway, Transcription Factors and Regulators | |
2735.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction