Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLI-2 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP223602H
Description
GLI-2 Polyclonal specifically detects GLI-2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
GLI-2 | |
Polyclonal | |
Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
P10070 | |
GLI2 | |
A synthetic peptide made to an internal portion of the human GLI-2 protein (between residues 300-400) [UniProt# P10070] | |
0.1 mL | |
Alzheimers Research, Cancer, Cell Cycle and Replication, Neuroscience, Stem Cell Signaling Pathway | |
2736 | |
Human, Mouse, Rat | |
IgG |
Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
HRP | |
PBS with No Preservative | |
GLI family zinc finger 2, GLI-Kruppel family member GLI2, glioma-associated oncogene family zinc finger 2, HPE9tax helper protein 1, oncogene GLI2, Tax helper protein, tax helper protein 2, tax-responsive element-2 holding protein, tax-responsive element-25-bp sequence binding protein, THP, THP1, THP2, zinc finger protein GLI2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human and mouse. Predicted to react with rat and hamster based on sequence homology. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction