Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLI-3 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP229627JF549
Description
GLI-3 Polyclonal specifically detects GLI-3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).Specifications
GLI-3 | |
Polyclonal | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular) | |
ACLSPAPAPHS, GCPS, GLI family zinc finger 3, GLI3 form of 190 kDa, GLI3 full length protein, GLI3-190, GLI3FL, GLI-Kruppel family member GLI3, glioma-associated oncogene family zinc finger 3, Greig cephalopolysyndactyly syndrome, oncogene GLI3, PAP-A, PAPA1, PAPBDNA-binding protein, PPDIV, zinc finger protein GLI3 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GLI3 | |
A synthetic peptide made to an internal portion of human Gli3 protein (between residues 50-120) [UniProt P10071] | |
0.1 mL | |
Cancer, Stem Cell Signaling Pathway | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction