Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLI-3 Rabbit anti-Human, Mouse, Rat, Clone: 8B1E2, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | GLI-3 |
---|---|
Clone | 8B1E2 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
GLI-3 Monoclonal antibody specifically detects GLI-3 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
GLI-3 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer, Stem Cell Signaling Pathway | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
2737 | |
IgG | |
Affinity purified |
8B1E2 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
ACLSPAPAPHS, GCPS, GLI family zinc finger 3, GLI3 form of 190 kDa, GLI3 full length protein, GLI3-190, GLI3FL, GLI-Kruppel family member GLI3, glioma-associated oncogene family zinc finger 3, Greig cephalopolysyndactyly syndrome, oncogene GLI3, PAP-A, PAPA1, PAPBDNA-binding protein, PPDIV, zinc finger protein GLI3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1301-1580 of human GLI-3 (P10071). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title