Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GLI1 Monoclonal Antibody (1B9F8)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA538530
Description
This antibody has been tested in indirect ELISA.
This gene encodes a member of the Kruppel family of zinc finger proteins. The encoded transcription factor is activated by the sonic hedgehog signal transduction cascade and regulates stem cell proliferation. The activity and nuclear localization of this protein is negatively regulated by p53 in an inhibitory loop. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
GLI1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P08151 | |
GLI1 | |
Purified recombinant fragment of human GLI1 (aa: 284-449) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1B9F8 | |
Unconjugated | |
GLI1 | |
AV235269; GLI; GLI family zinc finger 1; Gli1; GLI-Kruppel family member GLI1; Glioma-associated oncogene; glioma-associated oncogene 1; glioma-associated oncogene homolog; glioma-associated oncogene homolog 1 (zinc finger protein); oncogene GLI; Zfp5; Zfp-5; zinc finger protein 5; zinc finger protein GLI1 | |
Mouse | |
Protein G | |
RUO | |
2735 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction