Learn More
GLI1 Mouse anti-Human, Clone: OTI9A1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF804177

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
May regulate the transcription of specific genes during normal development. May play a role in craniofacial development and digital development, as well as development of the central nervous system and gastrointestinal tract. Mediates SHH signaling and thus cell proliferation and differentiation.Specifications
GLI1 | |
Monoclonal | |
Unconjugated | |
GLI1 | |
GLI | |
Mouse | |
Affinity Chromatography | |
RUO | |
2735 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI9A1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P08151 | |
GLI1 | |
Human recombit protein fragment corresponding to amino acids 1-410 of human GLI1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.