Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                GLI1 Polyclonal Antibody, Active Motif™
 
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Rabbit Polyclonal Antibody
$159.90 - $633.45
Specifications
| Antigen | GLI1 | 
|---|---|
| Concentration | Lot-Specific | 
| Applications | Western Blot | 
| Classification | Polyclonal | 
| Conjugate | Unconjugated | 
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
| 50-199-3243    | Active Motif 61216 | 10 μL | 
                                        
                                            
                                                
                                                
                                                
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $159.90
                                                 |  | |||||
| 50-199-3242    | Active Motif 61215 | 100 μL | 
                                        
                                            
                                                
                                                
                                                
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $633.45
                                                 |  | |||||
Description
This gene encodes a member of the Kruppel family of zinc finger proteins. The encoded transcription factor is activated by the sonic hedgehog signal transduction cascade and regulates stem cell proliferation. The activity and nuclear localization of this protein is negatively regulated by p53 in an inhibitory loop. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| GLI1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P08151 | |
| 2735 | |
| This GLI1 antibody was raised against a peptide within the C-terminal region of human GLI1. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| GLI1 | 
| Lot-Specific | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
| AV235269; Gli; GLI family zinc finger 1; GLI1; GLI-Kruppel family member GLI1; Glioma-associated oncogene; glioma-associated oncogene 1; Glioma-associated oncogene homolog; glioma-associated oncogene homolog 1 (zinc finger protein); Oncogene GLI; Zfp5; Zfp-5; zinc finger protein 5; Zinc finger protein GLI1 | |
| GLI1 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody | 
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
			
            