Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLI2 Rabbit anti-Mouse, Human, Polyclonal, MilliporeSigma™

Catalog No. ABN2241MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN2241MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN2241MI Supplier MilliporeSigma Supplier No. ABN2241
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-GLI2 is a highly specific rabbit polyclonal antibody that targets Zinc finger protein GLI2 and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLI2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Gli2,THP
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 12 amino acids from the internal edge of N-terminal region of Human Gli2.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_005261
Test Specificity This polyclonal antibody detects Gli2 in mouse brain. It targets an epitope within 12 amino acids from the N-terminal half of isoform 5.
Target Species Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.