Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLI4 Antibody (OTI3A9), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270833B
Description
GLI4 Monoclonal specifically detects GLI4 in Human samples. It is validated for Immunohistochemistry.Specifications
GLI4 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
GLI4 | |
Full length recombinant protein of human GLI4 (NP_612474) produced in E.coli. | |
0.1 mL | |
2738.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI3A9 | |
Immunohistochemistry | |
GLI family zinc finger 4, GLI-Kruppel family member GLI4, glioma-associated oncogene family zinc finger 4, HKR4GLI-Kruppel family member GLI4 (oncogene HKR4), Krueppel-related zinc finger protein 4, Protein HKR4, zinc finger protein GLI4, ZNF928 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction