Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLI4 Monoclonal Antibody (OTI3A9)
GREENER_CHOICE

Catalog No. PIMA526783
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526783 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526783 Supplier Invitrogen™ Supplier No. MA526783
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GLI4 Monoclonal Antibody for Western Blot, IHC (P)

GLI4 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLI4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3A9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Gli4
Gene Accession No. P10075
Gene Alias 9130023H24Rik; GLI family zinc finger 4; GLI4; GLI-Kruppel family member GLI4; GLI-Kruppel family member GLI4 (oncogene HKR4); glioma-associated oncogene family zinc finger 4; HKR4; Krueppel-related zinc finger protein 4; Protein HKR4; RIKEN cDNA 9130023H24 gene; zinc finger protein GLI4; ZNF928
Gene Symbols Gli4
Host Species Mouse
Immunogen Full length human recombinant protein of GLI4 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2738
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.